Search
Search

SLCO5A1| Gene Symbol: SLCO5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109226.2 | XP_013964701.1 | 6 - 7 | 2489 | 70 | |
| XM_022412334.1 | XP_022268042.1 | 7 - 8 | 2653 | 70 | ||
| XM_535098.6 | XP_535098.4 | 7 - 8 | 2648 | 70 | ||
Set Membership: |
|