Search
Search
| Gene Symbol: HSPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111037.2 | XP_013966512.1 | 6 - 7 | 591 | 100 | |
| XM_536018.6 | XP_536018.2 | 5 - 6 | 453 | 100 | ||
| XM_852308.5 | XP_857401.2 | 4 - 5 | 396 | 100 | ||
Set Membership: |
|