Search
Search

FILIP1L| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639388.3 | XP_005639445.1 | 889 | 93 | ||
| XM_022413927.1 | XP_022269635.1 | 541 | 93 | |||
| XM_022413928.1 | XP_022269636.1 | 318 | 93 | |||
| XM_022413929.1 | XP_022269637.1 | 313 | 93 | |||
| XM_022413930.1 | XP_022269638.1 | 193 | 93 | |||
| XM_022413931.1 | XP_022269639.1 | 179 | ||||
Set Membership: |
|