Search
Search
| Gene Symbol: VEZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639292.3 | XP_003639340.1 | 5 - 6 | 1151 | 61 | |
| XM_014116682.2 | XP_013972157.1 | 5 - 6 | 1169 | 61 | ||
| XM_014116685.2 | XP_013972160.1 | 6 - 7 | 1396 | 61 | ||
| XM_022423185.1 | XP_022278893.1 | 7 - 8 | 1519 | 61 | ||
| XM_022423186.1 | XP_022278894.1 | 7 - 8 | 1553 | 61 | ||
Set Membership: |
|