Search
Search
| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616548.3 | XP_005616605.1 | 3 - 4 | 1351 | 84 | |
| XM_005616549.3 | XP_005616606.1 | 3 - 4 | 1420 | 84 | ||
| XM_005616550.3 | XP_005616607.1 | 3 - 4 | 1322 | 84 | ||
| XM_541609.6 | XP_541609.1 | 3 - 4 | 1302 | 84 | ||
Set Membership: |
|