Search
Search
| Gene Symbol: CCDC30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119648.2 | XP_013975123.1 | 10 - 11 | 1651 | 83 | |
| XM_014119649.2 | XP_013975124.1 | 10 - 11 | 1571 | 83 | ||
| XM_014119650.2 | XP_013975125.1 | 10 - 11 | 1582 | 83 | ||
| XM_022427717.1 | XP_022283425.1 | 11 - 12 | 1708 | 83 | ||
| XM_022427718.1 | XP_022283426.1 | 11 - 12 | 1628 | 83 | ||
| XM_022427720.1 | XP_022283428.1 | 9 - 10 | 1312 | 83 | ||
| XM_022427721.1 | XP_022283429.1 | 8 - 9 | 1376 | 83 | ||
Set Membership: |
|