Search
Search
| Gene Symbol: DDX39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014377.1 | NP_001014399.1 | 5 - 6 | 730 | 79 | |
| XM_022425718.1 | XP_022281426.1 | 6 - 7 | 884 | 79 | ||
| XM_022425719.1 | XP_022281427.1 | 6 - 7 | 1002 | 79 | ||
| XM_022425720.1 | XP_022281428.1 | 6 - 7 | 884 | 79 | ||
Set Membership: |
|