Search
Search
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117533.2 | XP_013973008.1 | 1 - 2 | 231 | 65 | |
| XM_022424732.1 | XP_022280440.1 | 1 - 2 | 239 | 65 | ||
| XM_022424733.1 | XP_022280441.1 | 1 - 2 | 240 | 65 | ||
| XM_022424743.1 | XP_022280451.1 | 1 - 2 | 246 | 65 | ||
| XM_022424745.1 | XP_022280453.1 | 1 - 2 | 236 | 65 | ||
| XM_022424746.1 | XP_022280454.1 | 1 - 2 | 235 | 65 | ||
| XM_022424747.1 | XP_022280455.1 | 1 - 2 | 234 | 65 | ||
Set Membership: |
|