Search
Search
| Gene Symbol: RAD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632220.3 | XP_005632277.1 | 837 | 65 | ||
| XM_022406423.1 | XP_022262131.1 | 806 | 65 | |||
| XM_022406424.1 | XP_022262132.1 | 860 | 65 | |||
| XM_022406425.1 | XP_022262133.1 | 806 | 65 | |||
| XM_533750.6 | XP_533750.2 | 806 | 65 | |||
Set Membership: |
|