Search
Search
| Gene Symbol: CHKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121093.2 | XP_013976568.1 | 7 - 8 | 707 | 74 | |
| XM_014121094.1 | XP_013976569.1 | 6 - 7 | 554 | 74 | ||
| XM_014121095.2 | XP_013976570.1 | 7 - 8 | 673 | 74 | ||
| XM_014121097.2 | XP_013976572.1 | 7 - 8 | 638 | 74 | ||
| XM_022405582.1 | XP_022261290.1 | 8 - 9 | 897 | 74 | ||
| XM_022405584.1 | XP_022261292.1 | 8 - 9 | 902 | 74 | ||
| XM_022405585.1 | XP_022261293.1 | 7 - 8 | 841 | 74 | ||
| XM_022405586.1 | XP_022261294.1 | 8 - 9 | 703 | 74 | ||
| XM_533210.5 | XP_533210.3 | 7 - 8 | 608 | 74 | ||
Set Membership: |
|