Search
Search
| Gene Symbol: WDR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640555.3 | XP_005640612.1 | 8 - 9 | 887 | 66 | |
| XM_022415172.1 | XP_022270880.1 | 7 - 8 | 881 | 66 | ||
| XM_536036.6 | XP_536036.3 | 8 - 9 | 929 | 66 | ||
Set Membership: |
|