Search
Search
| Gene Symbol: SUMF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620921.3 | XP_005620978.1 | 679 | 73 | ||
| XM_014114144.2 | XP_013969619.1 | 513 | 73 | |||
| XM_022419521.1 | XP_022275229.1 | 735 | 73 | |||
| XM_022419522.1 | XP_022275230.1 | 692 | 73 | |||
Set Membership: |
|