Search
Search
| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419346.1 | XP_022275054.1 | 8 - 9 | 1144 | 68 | |
| XM_022419347.1 | XP_022275055.1 | 8 - 9 | 1010 | 68 | ||
| XM_022419348.1 | XP_022275056.1 | 8 - 9 | 1131 | 68 | ||
| XM_022419349.1 | XP_022275057.1 | 8 - 9 | 1145 | 68 | ||
| XM_022419350.1 | XP_022275058.1 | 7 - 8 | 1064 | 68 | ||
| XM_022419351.1 | XP_022275059.1 | 7 - 8 | 1051 | 68 | ||
| XM_022419352.1 | XP_022275060.1 | 6 - 7 | 781 | 68 | ||
| XM_022419353.1 | XP_022275061.1 | 6 - 7 | 1755 | 68 | ||
| XM_022419354.1 | XP_022275062.1 | 7 - 8 | 1100 | 68 | ||
| XM_022419355.1 | XP_022275063.1 | 6 - 7 | 1752 | 68 | ||
Set Membership: |
|