Search
Search
| Gene Symbol: VWA3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114556.2 | XP_013970031.1 | 18 - 19 | 1870 | 59 | |
| XM_022419963.1 | XP_022275671.1 | 18 - 19 | 1893 | 59 | ||
| XM_022419964.1 | XP_022275672.1 | 18 - 19 | 2028 | 59 | ||
| XM_022419965.1 | XP_022275673.1 | 18 - 19 | 2028 | 59 | ||
Set Membership: |
|