Search
Search

TBC1D32| Gene Symbol: TBC1D32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615601.3 | XP_005615658.2 | 3 - 4 | 458 | 153 | |
| XM_005615602.3 | XP_005615659.2 | 3 - 4 | 458 | 153 | ||
| XM_014113418.2 | XP_013968893.1 | 3 - 4 | 458 | 153 | ||
| XM_022419495.1 | XP_022275203.1 | 3 - 4 | 458 | 153 | ||
Set Membership: |
|