Search
Search
| Gene Symbol: PUS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626124.3 | XP_005626181.1 | 12 - 13 | 1450 | 70 | |
| XM_014117547.2 | XP_013973022.1 | 13 - 14 | 1686 | 70 | ||
| XM_014117548.2 | XP_013973023.1 | 14 - 15 | 2014 | 70 | ||
| XM_014117549.2 | XP_013973024.1 | 14 - 15 | 1977 | 70 | ||
| XM_022424773.1 | XP_022280481.1 | 13 - 14 | 1668 | 70 | ||
| XM_022424774.1 | XP_022280482.1 | 12 - 13 | 1369 | 70 | ||
| XM_022424775.1 | XP_022280483.1 | 12 - 13 | 1120 | 70 | ||
Set Membership: |
|