Search
Search
| Gene Symbol: MCF2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634114.3 | XP_005634171.1 | 3 - 4 | 833 | 70 | |
| XM_005634115.3 | XP_005634172.1 | 3 - 4 | 613 | 70 | ||
| XM_005634116.3 | XP_005634173.1 | 3 - 4 | 429 | 70 | ||
| XM_005634117.3 | XP_005634174.1 | 3 - 4 | 488 | 70 | ||
| XM_005634120.2 | XP_005634177.1 | 3 - 4 | 399 | 70 | ||
| XM_005634121.3 | XP_005634178.1 | 3 - 4 | 222 | 70 | ||
| XM_014106461.2 | XP_013961936.1 | 4 - 5 | 636 | 70 | ||
| XM_014106462.2 | XP_013961937.1 | 5 - 6 | 1147 | 70 | ||
| XM_014106464.2 | XP_013961939.1 | 5 - 6 | 1148 | 70 | ||
| XM_014106465.2 | XP_013961940.1 | 3 - 4 | 429 | 70 | ||
| XM_022408166.1 | XP_022263874.1 | 5 - 6 | 1147 | 70 | ||
| XM_022408167.1 | XP_022263875.1 | 5 - 6 | 1147 | 70 | ||
Set Membership: |
|