Search
Search
| Gene Symbol: FBXO40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639548.3 | XP_005639605.1 | 3 - 4 | 575 | 104 | |
| XM_022414009.1 | XP_022269717.1 | 4 - 5 | 258 | 104 | ||
| XM_022414010.1 | XP_022269718.1 | 3 - 4 | 309 | 104 | ||
Set Membership: |
|