Search
Search
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641067.1 | XP_005641124.1 | 6 - 7 | 850 | 85 | |
| XM_014111780.2 | XP_013967255.1 | 5 - 6 | 729 | 85 | ||
| XM_014111782.2 | XP_013967257.1 | 5 - 6 | 729 | 85 | ||
Set Membership: |
|