Search
Search
| Gene Symbol: RIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634721.3 | XP_005634778.1 | 12 - 13 | 2580 | 65 | |
| XM_005634722.3 | XP_005634779.1 | 11 - 12 | 2494 | 65 | ||
| XM_005634723.3 | XP_005634780.1 | 11 - 12 | 2433 | 65 | ||
| XM_005634725.2 | XP_005634782.1 | 9 - 10 | 2301 | 65 | ||
| XM_005634726.3 | XP_005634783.1 | 8 - 9 | 2273 | 65 | ||
| XM_014106942.2 | XP_013962417.1 | 10 - 11 | 2376 | 65 | ||
| XM_022409237.1 | XP_022264945.1 | 9 - 10 | 2262 | 65 | ||
| XM_022409238.1 | XP_022264946.1 | 11 - 12 | 2479 | 65 | ||
| XM_022409239.1 | XP_022264947.1 | 9 - 10 | 2302 | 65 | ||
Set Membership: |
|