Search
Search
| Gene Symbol: CEP78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615827.3 | XP_005615884.1 | 13 - 14 | 1503 | 130 | |
| XM_005615829.3 | XP_005615886.1 | 12 - 13 | 1481 | 130 | ||
| XM_014117446.2 | XP_013972921.1 | 13 - 14 | 1505 | 130 | ||
| XM_014117447.2 | XP_013972922.1 | 12 - 13 | 1483 | 130 | ||
| XM_022420889.1 | XP_022276597.1 | 11 - 12 | 1229 | 130 | ||
| XM_022420898.1 | XP_022276606.1 | 13 - 14 | 1504 | 130 | ||
| XM_022420901.1 | XP_022276609.1 | 11 - 12 | 1010 | 130 | ||
Set Membership: |
|