Search
Search
| Gene Symbol: NUSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638270.3 | XP_005638327.2 | 10 - 11 | 1191 | 74 | |
| XM_005638272.2 | XP_005638329.1 | 8 - 9 | 1132 | 74 | ||
| XM_005638273.3 | XP_005638330.2 | 10 - 11 | 1146 | 74 | ||
| XM_014109541.2 | XP_013965016.1 | 8 - 9 | 1088 | 74 | ||
| XM_022413051.1 | XP_022268759.1 | 8 - 9 | 1130 | 74 | ||
| XM_022413052.1 | XP_022268760.1 | 8 - 9 | 948 | 74 | ||
| XM_022413053.1 | XP_022268761.1 | 9 - 10 | 1247 | 74 | ||
| XM_022413054.1 | XP_022268762.1 | 9 - 10 | 1064 | 74 | ||
Set Membership: |
|