Search
Search
| Gene Symbol: CAMKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626071.3 | XP_005626128.1 | 2 - 3 | 575 | 130 | |
| XM_005626072.3 | XP_005626129.1 | 2 - 3 | 446 | 130 | ||
| XM_022424673.1 | XP_022280381.1 | 2 - 3 | 446 | 130 | ||
| XM_022424674.1 | XP_022280382.1 | 3 - 4 | 788 | 130 | ||
| XM_846713.5 | XP_851806.1 | 2 - 3 | 446 | 130 | ||
Set Membership: |
|