Search
Search
| Gene Symbol: SPG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638516.2 | XP_005638573.1 | 4 - 5 | 636 | 85 | |
| XM_005638518.3 | XP_005638575.1 | 4 - 5 | 581 | 85 | ||
| XM_014109515.2 | XP_013964990.1 | 4 - 5 | 641 | 85 | ||
| XM_014109517.2 | XP_013964992.1 | 3 - 4 | 407 | 85 | ||
| XM_022412793.1 | XP_022268501.1 | 3 - 4 | 329 | 85 | ||
| XM_022412794.1 | XP_022268502.1 | 3 - 4 | 343 | 85 | ||
| XM_847398.4 | XP_852491.3 | 3 - 4 | 403 | 85 | ||
Set Membership: |
|