Search
Search
| Gene Symbol: YPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636627.3 | XP_005636684.1 | 3 - 4 | 490 | 81 | |
| XM_022410816.1 | XP_022266524.1 | 3 - 4 | 501 | 81 | ||
| XM_022410817.1 | XP_022266525.1 | 3 - 4 | 430 | 81 | ||
Set Membership: |
|