Search
Search

PDCD1LG2| Gene Symbol: PDCD1LG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421848.1 | XP_022277556.1 | 6 - 7 | 1111 | 147 | |
| XM_022421859.1 | XP_022277567.1 | 6 - 7 | 963 | 147 | ||
| XM_847012.5 | XP_852105.3 | 6 - 7 | 1106 | 147 | ||
Set Membership: |
|