Search
Search

SLC18B1| Gene Symbol: SLC18B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615421.3 | XP_005615478.1 | 760 | 79 | ||
| XM_022417159.1 | XP_022272867.1 | 1081 | 79 | |||
| XM_022417166.1 | XP_022272874.1 | 936 | 79 | |||
| XM_848975.5 | XP_854068.2 | 904 | 79 | |||
Set Membership: |
|