Search
Search
| Gene Symbol: CCNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621685.3 | XP_005621742.1 | 16 - 17 | 1974 | 65 | |
| XM_022420238.1 | XP_022275946.1 | 9 - 10 | 1435 | 65 | ||
| XM_022420239.1 | XP_022275947.1 | 9 - 10 | 1347 | 65 | ||
Set Membership: |
|