Search
Search

DCUN1D4| Gene Symbol: DCUN1D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628180.3 | XP_005628237.2 | 2 - 3 | 270 | 101 | |
| XM_005628181.3 | XP_005628238.2 | 2 - 3 | 274 | 101 | ||
| XM_022426977.1 | XP_022282685.1 | 2 - 3 | 280 | 101 | ||
| XM_022426978.1 | XP_022282686.1 | 3 - 4 | 231 | 101 | ||
| XM_022426979.1 | XP_022282687.1 | 2 - 3 | 209 | 101 | ||
| XM_022426981.1 | XP_022282689.1 | 3 - 4 | 302 | 101 | ||
| XM_022426982.1 | XP_022282690.1 | 2 - 3 | 98 | 101 | ||
Set Membership: |
|