Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850028.5 | XP_855121.1 | 2 - 3 | 228 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |