Search
Search
| Gene Symbol: CDCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410959.1 | XP_022266667.1 | 4 - 5 | 936 | 67 | |
| XM_022410960.1 | XP_022266668.1 | 4 - 5 | 798 | 67 | ||
| XM_849304.5 | XP_854397.1 | 4 - 5 | 917 | 67 | ||
Set Membership: |
|