Search
Search
| Gene Symbol: TRDMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122442.2 | XP_013977917.1 | 1013 | 81 | ||
| XM_022407406.1 | XP_022263114.1 | 1078 | 81 | |||
| XM_022407410.1 | XP_022263118.1 | 757 | 81 | |||
| XM_843500.5 | XP_848593.1 | 990 | 81 | |||
Set Membership: |
|