Search
Search
| Gene Symbol: DYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623130.3 | XP_005623187.1 | 3 - 4 | 366 | 71 | |
| XM_022421670.1 | XP_022277378.1 | 3 - 4 | 517 | 71 | ||
| XM_022421671.1 | XP_022277379.1 | 3 - 4 | 514 | 71 | ||
| XM_537347.6 | XP_537347.2 | 3 - 4 | 510 | 71 | ||
Set Membership: |
|