Search
Search
| Gene Symbol: CALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629603.3 | XP_005629660.1 | 2004 | 58 | ||
| XM_014119846.2 | XP_013975321.1 | 1686 | 58 | |||
| XM_022403792.1 | XP_022259500.1 | 2632 | 58 | |||
| XM_022403793.1 | XP_022259501.1 | 2671 | 58 | |||
| XM_022403794.1 | XP_022259502.1 | 2674 | 58 | |||
| XM_022403795.1 | XP_022259503.1 | 2465 | 58 | |||
| XM_022403796.1 | XP_022259504.1 | 2632 | 58 | |||
Set Membership: |
|