Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Cf02672043_m1
          See other DZANK1 GE Assays ›
          Gene Symbol:
          DZANK1
          Gene Name:
          double zinc ribbon and ankyrin repeat domains 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.24: 4727996 - 4805799 on Build CanFam3.1
          Species:
          Dog
          Species Specific ID (Flybase ID):
          -
          Assay ID Cf02672043_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DZANK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_014107114.2 XP_013962589.1 20 - 21 2296 167
          XM_014107117.2 XP_013962592.1 20 - 21 2284 167
          XM_014107121.2 XP_013962596.1 20 - 21 2296 167
          XM_014107128.2 XP_013962603.1 19 - 20 2193 167
          XM_022409300.1 XP_022265008.1 20 - 21 2483 167
          XM_022409301.1 XP_022265009.1 20 - 21 2452 167
          XM_022409302.1 XP_022265010.1 19 - 20 2134 167
          XM_022409303.1 XP_022265011.1 20 - 21 2471 167
          XM_022409304.1 XP_022265012.1 20 - 21 2554 167
          XM_022409305.1 XP_022265013.1 20 - 21 2369 167
          XM_022409306.1 XP_022265014.1 20 - 21 2394 167
          XM_022409307.1 XP_022265015.1 20 - 21 2399 167
          XM_022409308.1 XP_022265016.1 20 - 21 2441 167
          XM_022409310.1 XP_022265018.1 19 - 20 2393 167
          XM_022409311.1 XP_022265019.1 19 - 20 2390 167
          XM_022409312.1 XP_022265020.1 19 - 20 2360 167
          XM_022409314.1 XP_022265022.1 19 - 20 2351 167
          XM_022409315.1 XP_022265023.1 19 - 20 2348 167
          XM_022409316.1 XP_022265024.1 18 - 19 2279 167
          XM_022409317.1 XP_022265025.1 18 - 19 2270 167
          XM_022409318.1 XP_022265026.1 18 - 19 2258 167
          XM_022409319.1 XP_022265027.1 18 - 19 2237 167
          XM_022409320.1 XP_022265028.1 20 - 21 2297 167
          XM_022409321.1 XP_022265029.1 20 - 21 2296 167
          XM_022409322.1 XP_022265030.1 17 - 18 2177 167
          XM_022409323.1 XP_022265031.1 19 - 20 2112 167
          XM_022409324.1 XP_022265032.1 14 - 15 1677 167
          XM_022409325.1 XP_022265033.1 14 - 15 1599 167

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline