Search
Search
| Gene Symbol: PDE8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416769.1 | XP_022272477.1 | 394 | 126 | ||
| XM_022416770.1 | XP_022272478.1 | 394 | 126 | |||
| XM_022416771.1 | XP_022272479.1 | 394 | 126 | |||
Set Membership: |
|