Search
Search

SULT1A1| Gene Symbol: SULT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003223.1 | NP_001003223.1 | 4 - 5 | 440 | 173 | |
| XM_014114387.1 | XP_013969862.1 | 4 - 5 | 721 | 173 | ||
| XM_022419477.1 | XP_022275185.1 | 4 - 5 | 651 | 173 | ||
| GenBank mRNA | AY069922.1 | - | 4 - 5 | 416 | 173 | |
| D29807.1 | - | 4 - 5 | 440 | 173 | ||
Set Membership: |
|