Search
Search
| Gene Symbol: DOCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637851.3 | XP_005637908.1 | 47 - 48 | 4851 | 139 | |
| XM_014108893.2 | XP_013964368.1 | 46 - 47 | 4887 | 139 | ||
| XM_022411679.1 | XP_022267387.1 | 47 - 48 | 4914 | 139 | ||
Set Membership: |
|