Search
Search
| Gene Symbol: FLI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619564.3 | XP_005619621.1 | 1120 | 111 | ||
| XM_005619566.3 | XP_005619623.1 | 887 | 111 | |||
| XM_022418094.1 | XP_022273802.1 | 882 | 111 | |||
| XM_546404.6 | XP_546404.2 | 1089 | 111 | |||
Set Membership: |
|