Search
Search
| Gene Symbol: NFKB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637669.3 | XP_005637726.1 | 17 - 18 | 2578 | 90 | |
| XM_005637671.3 | XP_005637728.1 | 17 - 18 | 2576 | 90 | ||
| XM_022412064.1 | XP_022267772.1 | 14 - 15 | 1795 | 90 | ||
Set Membership: |
|