Search
Search
| Gene Symbol: PARG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637487.3 | XP_005637544.1 | 6 - 7 | 1622 | 147 | |
| XM_005637488.3 | XP_005637545.1 | 6 - 7 | 1550 | 147 | ||
| XM_005637489.3 | XP_005637546.1 | 6 - 7 | 1537 | 147 | ||
| XM_022411745.1 | XP_022267453.1 | 5 - 6 | 1403 | 147 | ||
| XM_534946.6 | XP_534946.2 | 6 - 7 | 1719 | 147 | ||
Set Membership: |
|