Search
Search
| Gene Symbol: NRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638876.4 | XP_003638924.1 | 895 | 108 | ||
| XM_005616946.3 | XP_005617003.1 | 895 | 108 | |||
| XM_005616947.3 | XP_005617004.1 | 895 | 108 | |||
| XM_535142.6 | XP_535142.3 | 895 | 108 | |||
Set Membership: |
|