Search
Search
| Gene Symbol: LAMA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435003.4 | XP_003435051.1 | 7 - 8 | 1624 | 119 | |
| XM_537297.6 | XP_537297.2 | 44 - 45 | 5634 | 119 | ||
| GenBank mRNA | AF236865.1 | - | 7 - 8 | 783 | 119 | |
Set Membership: |
|