Search
Search
| Gene Symbol: USP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638495.3 | XP_005638552.1 | 14 - 15 | 1536 | 177 | |
| XM_005638496.2 | XP_005638553.1 | 15 - 16 | 1526 | 177 | ||
| XM_005638497.3 | XP_005638554.1 | 15 - 16 | 1540 | 177 | ||
| XM_022412740.1 | XP_022268448.1 | 14 - 15 | 1422 | 177 | ||
| XM_544715.6 | XP_544715.3 | 13 - 14 | 1488 | 177 | ||
Set Membership: |
|