Search
Search
| Gene Symbol: STAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641778.3 | XP_005641835.1 | 18 - 19 | 2254 | 130 | |
| XM_005641779.3 | XP_005641836.1 | 18 - 19 | 2030 | 130 | ||
| XM_005641780.2 | XP_005641837.1 | 18 - 19 | 1951 | 130 | ||
| XM_022416331.1 | XP_022272039.1 | 17 - 18 | 1827 | 130 | ||
| XM_022416332.1 | XP_022272040.1 | 17 - 18 | 1827 | 130 | ||
| XM_022416333.1 | XP_022272041.1 | 16 - 17 | 1848 | 130 | ||
| XM_022416334.1 | XP_022272042.1 | 14 - 15 | 1527 | 130 | ||
| XM_859783.5 | XP_864876.2 | 19 - 20 | 2182 | 130 | ||
Set Membership: |
|