Search
Search
| Gene Symbol: ITCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634950.3 | XP_005635007.1 | 21 - 22 | 2450 | 145 | |
| XM_014107005.2 | XP_013962480.1 | 20 - 21 | 2256 | 145 | ||
| XM_022409130.1 | XP_022264838.1 | 20 - 21 | 2332 | 145 | ||
| XM_022409131.1 | XP_022264839.1 | 18 - 19 | 2089 | 145 | ||
| XM_022409132.1 | XP_022264840.1 | 15 - 16 | 1827 | 145 | ||
| XM_846367.5 | XP_851460.1 | 21 - 22 | 2450 | 145 | ||
Set Membership: |
|