Search
Search
| Gene Symbol: MAP2K6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624291.2 | XP_005624348.2 | 699 | 130 | ||
| XM_005624292.3 | XP_005624349.1 | 637 | 130 | |||
| XM_022422869.1 | XP_022278577.1 | 699 | 130 | |||
| XM_022422870.1 | XP_022278578.1 | 778 | 130 | |||
| XM_022422871.1 | XP_022278579.1 | 669 | 130 | |||
| XM_845930.5 | XP_851023.1 | 935 | 130 | |||
Set Membership: |
|