Search
Search

PLEKHG5| Gene Symbol: PLEKHG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620459.3 | XP_005620516.1 | 1984 | 152 | ||
| XM_005620463.3 | XP_005620520.1 | 1982 | 152 | |||
| XM_022418986.1 | XP_022274694.1 | 1935 | 152 | |||
| XM_022418987.1 | XP_022274695.1 | 1857 | 152 | |||
Set Membership: |
|