Search
Search

C21H11orf16| Gene Symbol: C21H11orf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122607.2 | XP_013978082.1 | 5 | 1000 | 175 | |
| XM_022407818.1 | XP_022263526.1 | 5 | 1005 | 175 | ||
| XM_022407819.1 | XP_022263527.1 | 5 | 996 | 175 | ||
Set Membership: |
|