Search
Search

CTDSPL2| Gene Symbol: CTDSPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638357.3 | XP_005638414.1 | 10 - 11 | 1318 | 142 | |
| XM_005638359.3 | XP_005638416.1 | 9 - 10 | 1256 | 142 | ||
| XM_005638360.3 | XP_005638417.1 | 9 - 10 | 1074 | 142 | ||
| XM_005638361.3 | XP_005638418.1 | 9 - 10 | 1262 | 142 | ||
| XM_022412542.1 | XP_022268250.1 | 9 - 10 | 1153 | 142 | ||
| XM_855561.5 | XP_860654.1 | 9 - 10 | 1260 | 142 | ||
Set Membership: |
|